Search
Search

pitr-1| Gene Symbol: pitr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380320.1 | NP_001367368.1 | 5 - 6 | 656 | 73 | |
| NM_001380321.1 | NP_001368450.1 | 3 - 4 | 440 | 73 | ||
| NR_131566.1 | 5 - 6 | 644 | 73 | |||
Set Membership: |
|