Search
Search

daf-15| Gene Symbol: daf-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306928.3 | NP_001293857.1 | 7 - 8 | 1383 | 89 | |
| NM_001306929.3 | NP_001293858.1 | 7 - 8 | 1390 | 89 | ||
| NM_001380409.1 | NP_001368188.1 | 8 - 9 | 1439 | 89 | ||
| NM_001380410.1 | NP_001366749.1 | 8 - 9 | 1439 | 89 | ||
| NM_069252.7 | NP_501653.1 | 7 - 8 | 1381 | 89 | ||
| GenBank mRNA | AY396716.1 | - | 8 - 9 | 1439 | 89 | |
Set Membership: |
|