Search
Search

| Gene Symbol: wrt-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383164.1 | 4 - 5 | 610 | 145 | ||
| Gene Symbol: cng-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028023.3 | NP_001023194.1 | 4 - 5 | 638 | 145 | |
Set Membership: |
|