Search
Search

fbl-1| Gene Symbol: fbl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028066.6 | NP_001023237.1 | 13 - 14 | 1946 | 91 | |
| NM_001306947.3 | NP_001293876.1 | 11 - 12 | 1794 | 91 | ||
| NM_001306948.3 | NP_001293877.1 | 11 - 12 | 1677 | 91 | ||
| NM_069293.6 | NP_501694.2 | 13 - 14 | 2092 | 91 | ||
| GenBank mRNA | AF051401.1 | - | 13 - 14 | 1929 | 91 | |
| AY851363.1 | - | 13 - 14 | 2046 | 91 | ||
Set Membership: |
|