Search
Search

| Gene Symbol: ndx-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136381.4 | NP_001129853.1 | 4 - 5 | 657 | 87 | |
| NM_069650.6 | NP_502051.2 | 5 - 6 | 754 | 87 | ||
| GenBank mRNA | AF305937.1 | - | 4 - 5 | 657 | 87 | |
| Gene Symbol: F13H10.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171417.7 | NP_741486.1 | 15 - 16 | 3168 | 87 | |
Set Membership: |
|