Search
Search

aptf-2| Gene Symbol: aptf-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268811.3 | NP_001255740.1 | 2 - 3 | 606 | 95 | |
| NM_001268812.3 | NP_001255741.1 | 1 - 2 | 497 | 95 | ||
Set Membership: |
|