Search
Search

nhr-5| Gene Symbol: nhr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276894.4 | NP_001263823.2 | 4 - 5 | 391 | 72 | |
| NM_001383253.1 | 4 - 5 | 388 | 72 | |||
| NR_101983.1 | 4 - 5 | 322 | 72 | |||
Set Membership: |
|