Search
Search

srbc-20| Gene Symbol: srbc-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313133.3 | NP_001300062.1 | 2 - 3 | 414 | 151 | |
| NM_071179.4 | NP_503580.2 | 3 - 4 | 522 | 151 | ||
| NR_132529.1 | 2 - 3 | 252 | 151 | |||
Set Membership: |
|