Search
Search

| Gene Symbol: nhr-219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322579.3 | NP_001309650.1 | 3 - 4 | 658 | 132 | |
| Gene Symbol: nhr-44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072912.7 | NP_505313.1 | 3 - 4 | 657 | 132 | |
Set Membership: |
|