Search
Search

ppm-1.A| Gene Symbol: ppm-1.A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028671.4 | NP_001023842.1 | 3 - 4 | 780 | 91 | |
| NM_001028672.7 | NP_001023843.1 | 2 - 3 | 220 | 91 | ||
| NM_001129457.4 | NP_001122929.1 | 2 - 3 | 127 | 91 | ||
Set Membership: |
|