Search
Search

fln-2| Gene Symbol: fln-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270145.4 | NP_001257074.2 | 24 - 25 | 5996 | 81 | |
| NM_001313392.3 | NP_001300321.1 | 13 | 4397 | 81 | ||
| NM_001313394.3 | NP_001300323.1 | 31 - 32 | 6785 | 81 | ||
| NM_001313396.3 | NP_001300325.1 | 26 - 27 | 6260 | 81 | ||
| NM_001313398.3 | NP_001300327.1 | 23 - 24 | 5828 | 81 | ||
| NM_001313400.3 | NP_001300329.1 | 21 - 22 | 5636 | 81 | ||
| NM_001313402.3 | NP_001300331.1 | 16 - 17 | 4913 | 81 | ||
| NM_001373422.1 | NP_001359882.1 | 42 - 43 | 8552 | 81 | ||
| NM_001373424.2 | NP_001360664.1 | 41 | 8408 | 81 | ||
| NM_001373426.2 | NP_001360665.1 | 41 | 8417 | 81 | ||
| NM_001373428.2 | NP_001360667.1 | 40 | 8324 | 81 | ||
| NR_132548.1 | 43 - 44 | 9005 | 81 | |||
Set Membership: |
|