Search
Search

pgp-5| Gene Symbol: pgp-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270187.3 | NP_001257116.1 | 8 - 9 | 1127 | 154 | |
| NM_001270188.3 | NP_001257117.1 | 5 - 6 | 784 | 154 | ||
Set Membership: |
|