Search
Search

lgc-29| Gene Symbol: lgc-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038424.4 | NP_001033513.2 | 7 - 8 | 705 | 84 | |
| NM_001330830.1 | NP_001317792.1 | 7 - 8 | 659 | 84 | ||
Set Membership: |
|