Search
Search
| Gene Symbol: EMSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407659.1 | XP_022263367.1 | 5 - 6 | 410 | 120 | |
| XM_022407662.1 | XP_022263370.1 | 5 - 6 | 410 | 120 | ||
| XM_022407663.1 | XP_022263371.1 | 5 - 6 | 452 | 120 | ||
| XM_022407664.1 | XP_022263372.1 | 5 - 6 | 409 | 120 | ||
Set Membership: |
|