Search
Search
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003183.1 | NP_001003183.1 | 2 - 3 | 241 | 70 | |
| GenBank mRNA | AB191461.1 | - | 2 - 3 | 271 | 70 | |
| AF045773.1 | - | 2 - 3 | 241 | 70 | ||
| U96127.1 | - | 2 - 3 | 186 | 70 | ||
Set Membership: |
|