Search
Search
| Gene Symbol: GPX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115119.1 | NP_001108591.1 | 1 - 2 | 266 | 66 | |
| GenBank mRNA | AY572225.1 | - | 1 - 2 | 206 | 66 | |
| DN270560.1 | - | 1 - 2 | 308 | 66 | ||
| DN870414.1 | - | 1 - 2 | 182 | 66 | ||
Set Membership: |
|