Search
Search
| Gene Symbol: SEMA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622707.3 | XP_005622764.3 | 11 - 12 | 1458 | 63 | |
| XM_014115323.2 | XP_013970798.1 | 11 - 12 | 1458 | 63 | ||
| XM_014115325.2 | XP_013970800.1 | 11 - 12 | 1435 | 63 | ||
| XM_014115326.2 | XP_013970801.1 | 11 - 12 | 1410 | 63 | ||
| XM_014115327.2 | XP_013970802.1 | 11 - 12 | 1529 | 63 | ||
| XM_014115328.1 | XP_013970803.1 | 10 - 11 | 1284 | 63 | ||
| XM_014115329.1 | XP_013970804.1 | 10 - 11 | 1257 | 63 | ||
| XM_022421269.1 | XP_022276977.1 | 10 - 11 | 1359 | 63 | ||
| XM_022421270.1 | XP_022276978.1 | 10 - 11 | 1359 | 63 | ||
| XM_022421271.1 | XP_022276979.1 | 11 - 12 | 3116 | 63 | ||
Set Membership: |
|