Search
Search
| Gene Symbol: STK36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640657.3 | XP_005640714.1 | 16 - 17 | 2270 | 89 | |
| XM_005640658.3 | XP_005640715.1 | 16 - 17 | 2207 | 89 | ||
| XM_005640660.3 | XP_005640717.1 | 16 - 17 | 2147 | 89 | ||
| XM_005640661.3 | XP_005640718.1 | 15 - 16 | 2034 | 89 | ||
| XM_005640663.3 | XP_005640720.1 | 12 - 13 | 1575 | 89 | ||
| XM_022415075.1 | XP_022270783.1 | 16 - 17 | 2194 | 89 | ||
| XM_022415076.1 | XP_022270784.1 | 13 - 14 | 1962 | 89 | ||
| XM_536072.6 | XP_536072.3 | 16 - 17 | 2223 | 89 | ||
Set Membership: |
|