Search
Search
| Gene Symbol: AAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635002.3 | XP_005635059.1 | 3 - 4 | 1611 | 91 | |
| XM_022409009.1 | XP_022264717.1 | 4 - 5 | 1272 | 91 | ||
| XM_022409010.1 | XP_022264718.1 | 4 - 5 | 1787 | 91 | ||
| XM_022409011.1 | XP_022264719.1 | 4 - 5 | 1786 | 91 | ||
| XM_022409012.1 | XP_022264720.1 | 4 - 5 | 1892 | 91 | ||
| XM_534409.6 | XP_534409.1 | 3 - 4 | 1161 | 91 | ||
Set Membership: |
|