Search
Search

SLC25A23| Gene Symbol: SLC25A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632988.1 | XP_005633045.1 | 4 - 5 | 580 | 101 | |
| XM_005632989.3 | XP_005633046.1 | 4 - 5 | 580 | 101 | ||
| XM_014121925.1 | XP_013977400.1 | 4 - 5 | 580 | 101 | ||
| XM_014121926.2 | XP_013977401.1 | 4 - 5 | 580 | 101 | ||
Set Membership: |
|