Search
Search

DPY19L2| Gene Symbol: DPY19L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022427573.1 | XP_022283281.1 | 12 - 13 | 1466 | 103 | |
| XM_532514.5 | XP_532514.3 | 12 - 13 | 1320 | 103 | ||
Set Membership: |
|