Search
Search
| Gene Symbol: AUH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615952.3 | XP_005616009.1 | 2 - 3 | 369 | 132 | |
| XM_022422006.1 | XP_022277714.1 | 2 - 3 | 371 | 132 | ||
| XM_022422009.1 | XP_022277717.1 | 2 - 3 | 178 | 132 | ||
| XM_533549.6 | XP_533549.2 | 2 - 3 | 369 | 132 | ||
Set Membership: |
|