Search
Search

CFAP221| Gene Symbol: CFAP221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631908.1 | XP_005631965.1 | 22 - 23 | 2496 | 86 | |
| XM_005631909.2 | XP_005631966.1 | 21 - 22 | 2414 | 86 | ||
| XM_022406055.1 | XP_022261763.1 | 22 - 23 | 2497 | 86 | ||
| XM_022406056.1 | XP_022261764.1 | 22 - 23 | 2502 | 86 | ||
| XM_022406057.1 | XP_022261765.1 | 22 - 23 | 2499 | 86 | ||
| XM_022406058.1 | XP_022261766.1 | 22 - 23 | 2561 | 86 | ||
| XM_022406059.1 | XP_022261767.1 | 22 - 23 | 2522 | 86 | ||
| XM_022406060.1 | XP_022261768.1 | 22 - 23 | 2510 | 86 | ||
| XM_022406061.1 | XP_022261769.1 | 21 - 22 | 2415 | 86 | ||
Set Membership: |
|