Search
Search
| Gene Symbol: NHSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615453.3 | XP_005615510.1 | 4850 | 61 | ||
| XM_005615454.3 | XP_005615511.1 | 4718 | 61 | |||
| XM_005615455.3 | XP_005615512.1 | 4277 | 61 | |||
| XM_014112351.2 | XP_013967826.1 | 4569 | 61 | |||
| XM_022417475.1 | XP_022273183.1 | 4411 | 61 | |||
| XM_022417481.1 | XP_022273189.1 | 4510 | 61 | |||
Set Membership: |
|