Search
Search
| Gene Symbol: ACOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618490.3 | XP_005618547.1 | 17 - 18 | 2227 | 63 | |
| XM_005618491.3 | XP_005618548.1 | 17 - 18 | 2174 | 63 | ||
| XM_005618492.3 | XP_005618549.1 | 17 - 18 | 2104 | 63 | ||
Set Membership: |
|