Search
Search
| Gene Symbol: DPYSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617361.2 | XP_005617418.1 | 13 - 14 | 1637 | 79 | |
| XM_005617362.3 | XP_005617419.1 | 13 - 14 | 1610 | 79 | ||
| XM_014108278.2 | XP_013963753.1 | 13 - 14 | 1652 | 79 | ||
| XM_544332.6 | XP_544332.3 | 13 - 14 | 1766 | 79 | ||
Set Membership: |
|