Search
Search

SLC39A12| Gene Symbol: SLC39A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014122422.2 | XP_013977897.1 | 1729 | 71 | ||
| XM_022407364.1 | XP_022263072.1 | 1841 | 71 | |||
| XM_022407372.1 | XP_022263080.1 | 1577 | 71 | |||
| XM_843257.5 | XP_848350.2 | 1843 | 71 | |||
Set Membership: |
|