Search
Search
| Gene Symbol: RNPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621851.3 | XP_005621908.1 | 1 - 2 | 191 | 127 | |
| XM_014114797.2 | XP_013970272.1 | 1 - 2 | 191 | 127 | ||
| XM_014114798.2 | XP_013970273.1 | 1 - 2 | 191 | 127 | ||
| XM_022420407.1 | XP_022276115.1 | 1 - 2 | 191 | 127 | ||
| XM_022420408.1 | XP_022276116.1 | 1 - 2 | 307 | 127 | ||
| XM_022420409.1 | XP_022276117.1 | 1 - 2 | 191 | 127 | ||
| XM_022420410.1 | XP_022276118.1 | 1 - 2 | 191 | 127 | ||
Set Membership: |
|