Search
Search
| Gene Symbol: SNED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635965.2 | XP_005636022.1 | 6 - 7 | 1059 | 81 | |
| XM_022410064.1 | XP_022265772.1 | 6 - 7 | 1059 | 81 | ||
| XM_022410065.1 | XP_022265773.1 | 6 - 7 | 1059 | 81 | ||
| XM_022410066.1 | XP_022265774.1 | 6 - 7 | 1059 | 81 | ||
| XM_022410067.1 | XP_022265775.1 | 6 - 7 | 1059 | 81 | ||
| XM_022410068.1 | XP_022265776.1 | 5 - 6 | 771 | 81 | ||
| XM_022410069.1 | XP_022265777.1 | 6 - 7 | 755 | 81 | ||
Set Membership: |
|