Search
Search
| Gene Symbol: EHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423499.1 | XP_022279207.1 | 14 - 15 | 2365 | 63 | |
| XM_022423501.1 | XP_022279209.1 | 14 - 15 | 2344 | 63 | ||
| XM_022423502.1 | XP_022279210.1 | 14 - 15 | 2341 | 63 | ||
Set Membership: |
|