Search
Search

DENND1A| Gene Symbol: DENND1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625388.3 | XP_005625445.1 | 11 - 12 | 1451 | 134 | |
| XM_005625389.3 | XP_005625446.1 | 11 - 12 | 1450 | 134 | ||
| XM_014117075.2 | XP_013972550.1 | 11 - 12 | 1452 | 134 | ||
| XM_022423856.1 | XP_022279564.1 | 12 - 13 | 1575 | 134 | ||
| XM_022423857.1 | XP_022279565.1 | 12 - 13 | 1575 | 134 | ||
| XM_022423858.1 | XP_022279566.1 | 12 - 13 | 1489 | 134 | ||
| XM_022423859.1 | XP_022279567.1 | 12 - 13 | 1575 | 134 | ||
| XM_022423860.1 | XP_022279568.1 | 12 - 13 | 1023 | 134 | ||
| XM_022423861.1 | XP_022279569.1 | 11 - 12 | 1419 | 134 | ||
| XM_022423862.1 | XP_022279570.1 | 11 - 12 | 1413 | 134 | ||
| XM_022423863.1 | XP_022279571.1 | 12 - 13 | 1574 | 134 | ||
| XM_022423864.1 | XP_022279572.1 | 11 - 12 | 846 | 134 | ||
| XM_022423865.1 | XP_022279573.1 | 11 - 12 | 947 | 134 | ||
| XM_022423866.1 | XP_022279574.1 | 4 - 5 | 347 | 134 | ||
Set Membership: |
|