Search
Search
| Gene Symbol: MTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641934.3 | XP_005641991.1 | 6 - 7 | 578 | 94 | |
| XM_005641935.3 | XP_005641992.1 | 5 - 6 | 420 | 94 | ||
| XM_005641936.2 | XP_005641993.1 | 2 - 3 | 176 | 94 | ||
| XM_850116.5 | XP_855209.1 | 5 - 6 | 420 | 94 | ||
Set Membership: |
|