Search
Search
| Gene Symbol: ATRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641515.3 | XP_005641572.1 | 15 - 16 | 4776 | 82 | |
| XM_005641516.3 | XP_005641573.1 | 14 - 15 | 4662 | 82 | ||
| XM_014111550.2 | XP_013967025.1 | 14 - 15 | 4666 | 82 | ||
| XM_538084.6 | XP_538084.2 | 15 - 16 | 4743 | 82 | ||
| XM_855031.5 | XP_860124.2 | 14 - 15 | 4665 | 82 | ||
Set Membership: |
|