Search
Search
| Gene Symbol: ABCG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413077.1 | XP_022268785.1 | 7 - 8 | 945 | 163 | |
| XM_022413078.1 | XP_022268786.1 | 7 - 8 | 945 | 163 | ||
| XM_022413079.1 | XP_022268787.1 | 7 - 8 | 853 | 163 | ||
Set Membership: |
|