Search
Search
| Gene Symbol: CDK16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641272.3 | XP_005641329.1 | 5 - 6 | 813 | 85 | |
| XM_005641273.3 | XP_005641330.1 | 5 - 6 | 812 | 85 | ||
| XM_005641274.3 | XP_005641331.1 | 5 - 6 | 930 | 85 | ||
| XM_005641276.2 | XP_005641333.1 | 5 - 6 | 913 | 85 | ||
| XM_022416226.1 | XP_022271934.1 | 5 - 6 | 546 | 85 | ||
| XM_538015.6 | XP_538015.3 | 5 - 6 | 928 | 85 | ||
Set Membership: |
|