Search
Search
| Gene Symbol: NPHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616692.3 | XP_005616749.1 | 7 - 8 | 1284 | 105 | |
| XM_022403537.1 | XP_022259245.1 | 7 - 8 | 1284 | 105 | ||
| XM_022403540.1 | XP_022259248.1 | 4 - 5 | 432 | 105 | ||
| XM_541685.6 | XP_541685.3 | 7 - 8 | 1283 | 105 | ||
Set Membership: |
|