Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107559.2 | XP_013963034.1 | 4 | 1329 | 138 | |
XM_543225.6 | XP_543225.4 | 5 | 1259 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |