Search
Search

CYP39A1| Gene Symbol: CYP39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627418.3 | XP_005627475.1 | 10 - 11 | 1501 | 132 | |
| XM_005627419.2 | XP_005627476.1 | 11 - 12 | 1318 | 132 | ||
| XM_014118490.2 | XP_013973965.1 | 9 - 10 | 1127 | 132 | ||
| XM_014118491.1 | XP_013973966.1 | 8 - 9 | 992 | 132 | ||
| XM_022426122.1 | XP_022281830.1 | 9 - 10 | 1014 | 132 | ||
| XM_538946.6 | XP_538946.2 | 9 - 10 | 1366 | 132 | ||
Set Membership: |
|