Search
Search
| Gene Symbol: PAX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626823.3 | XP_005626880.2 | 10 | 2087 | 91 | |
| XM_014117863.2 | XP_013973338.1 | 9 | 2030 | 91 | ||
| XM_022425611.1 | XP_022281319.1 | 10 | 1219 | 91 | ||
| XM_022425613.1 | XP_022281321.1 | 10 | 2057 | 91 | ||
| XM_849748.5 | XP_854841.2 | 9 | 1162 | 91 | ||
Set Membership: |
|