Search
Search
| Gene Symbol: TGIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635003.2 | XP_005635060.1 | 2 | 265 | 99 | |
| XM_005635004.3 | XP_005635061.1 | 2 | 285 | 99 | ||
| XM_005635005.2 | XP_005635062.1 | 2 | 207 | 99 | ||
| XM_005635006.3 | XP_005635063.1 | 2 | 348 | 99 | ||
| XM_005635007.3 | XP_005635064.1 | 2 | 260 | 99 | ||
| XM_014106903.2 | XP_013962378.1 | 2 | 263 | 99 | ||
| XM_860525.5 | XP_865618.1 | 2 | 269 | 99 | ||
Set Membership: |
|