Search
Search
| Gene Symbol: HYAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632508.3 | XP_005632565.1 | 2 | 2827 | 108 | |
| XM_022406690.1 | XP_022262398.1 | 1 | 1248 | 108 | ||
| XM_845674.5 | XP_850767.3 | 1 | 1246 | 108 | ||
Set Membership: |
|