Search
Search
| Gene Symbol: FAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637014.3 | XP_005637071.1 | 13 - 14 | 1758 | 135 | |
| XM_005637015.3 | XP_005637072.1 | 12 - 13 | 1688 | 135 | ||
| XM_022411381.1 | XP_022267089.1 | 12 - 13 | 1599 | 135 | ||
| XM_022411382.1 | XP_022267090.1 | 11 - 12 | 1454 | 135 | ||
| XM_022411383.1 | XP_022267091.1 | 11 - 12 | 1568 | 135 | ||
| XM_022411384.1 | XP_022267092.1 | 11 - 12 | 1655 | 135 | ||
| XM_022411385.1 | XP_022267093.1 | 11 - 12 | 1602 | 135 | ||
| XM_022411386.1 | XP_022267094.1 | 10 - 11 | 1392 | 135 | ||
Set Membership: |
|