Search
Search
| Gene Symbol: WWC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434507.4 | XP_003434555.1 | 6 - 7 | 1068 | 148 | |
| XM_005619227.3 | XP_005619284.1 | 6 - 7 | 1068 | 148 | ||
| XM_014113022.2 | XP_013968497.1 | 6 - 7 | 1068 | 148 | ||
| XM_014113023.2 | XP_013968498.1 | 6 - 7 | 1068 | 148 | ||
| XM_014113024.2 | XP_013968499.1 | 6 - 7 | 1071 | 148 | ||
| XM_022417719.1 | XP_022273427.1 | 6 - 7 | 688 | 148 | ||
| XM_536435.6 | XP_536435.2 | 6 - 7 | 1068 | 148 | ||
Set Membership: |
|