Search
Search

Guca2a| Gene Symbol: Guca2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501825.2 | XP_003501873.2 | 2 - 3 | 552 | 79 | |
| XM_007642474.1 | XP_007640664.1 | 1 - 2 | 868 | 79 | ||
Set Membership: |
|