Search
Search

Ndst2| Gene Symbol: Ndst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652664.1 | XP_007650854.1 | 3 - 4 | 1684 | 61 | |
| GenBank mRNA | JI865790.1 | - | 2 - 3 | 294 | 61 | |
| JP055354.1 | - | 2 - 3 | 294 | 61 | ||
Set Membership: |
|