Search
Search

Ppp2r3c| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513764.2 | XP_003513812.1 | 10 - 11 | 1027 | 69 | |
| GenBank mRNA | JI872651.1 | - | 9 - 10 | 919 | 69 | |
| JP046019.1 | - | 9 - 10 | 919 | 69 | ||
Set Membership: |
|