Search
Search

Tmem164| Gene Symbol: Tmem164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649379.1 | XP_007647569.1 | 5 - 6 | 386 | 64 | |
| XM_007649380.1 | XP_007647570.1 | 6 - 7 | 479 | 64 | ||
Set Membership: |
|