Search
Search

Znf76| Gene Symbol: Znf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506290.2 | XP_003506338.1 | 9 - 10 | 1272 | 67 | |
| XM_007646796.1 | XP_007644986.1 | 9 - 10 | 1273 | 67 | ||
| GenBank mRNA | JI877658.1 | - | 9 - 10 | 1197 | 67 | |
| JP051495.1 | - | 9 - 10 | 1197 | 67 | ||
Set Membership: |
|