Search
Search

Tank| Gene Symbol: Tank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509037.2 | XP_003509085.1 | 2 - 3 | 272 | 83 | |
| GenBank mRNA | JI871967.1 | - | 2 - 3 | 207 | 83 | |
| JP045767.1 | - | 2 - 3 | 203 | 83 | ||
Set Membership: |
|