Search
Search

Immp1l| Gene Symbol: Immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497564.2 | XP_003497612.1 | 4 - 5 | 472 | 101 | |
| XM_007613937.1 | XP_007612127.1 | 4 - 5 | 510 | 101 | ||
| XM_007613952.1 | XP_007612142.1 | 5 - 6 | 766 | 101 | ||
| XM_007613960.1 | XP_007612150.1 | 5 - 6 | 728 | 101 | ||
Set Membership: |
|